Would you like to log in again?
Methyl Green-Pyronin Y solution stain for histology
Supplier: Epredia
Synonyms: MGP
87011 87011
84000-302EA 486.21 USD InStock
CA84000-302 84000-302
Methyl Green-Pyronin Y solution stain for histology
Methyl Green-Pyronin Y
Concentrated solution for Identification of Plasma Cells and RNA in Tissue Sections
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty