Would you like to log in again?
Temperature and Humidity Controlled Glove Box, Plas-Labs™
Supplier: Plas-Labs
890-THC/DT 890-THC 890-THC/EXP
87000-728EA 40745.38 USD InStock
CA87000-728 87000-728 87000-730
Temperature and Humidity Controlled Glove Box, Plas-Labs™
Glove Boxes and Glove Bags
For laboratory isolation applications that require the maintenance of accurate temperature and humidity levels.
Roomy, single-station glove box includes a vacuum-capable transfer chamber, Hypalon® gloves, large full-color display, inert gas hook up, internal 110V power, and a serial connection interface for datalogging. The chamber has a one-piece clear acrylic top and die-molded bottom. Easy-to-use touch screen display features password protection; four preset conditions; chamber-temperature, room-temperature, and humidity readings; date and time display; and four selectable high/low alarms. Two inlet/outlet gas valves are located on the main chamber and on the transfer chamber. Units include a relative humidity transmitter with a 244cm (8') sensor cable, a platinum RTD with sealing gland, a nebulizer, and a thermoelectric assembly (Peltier device). Unit 97000-730 includes a drying train package with two molecular sieve canisters and circulation pump.
UL listed. CE marked.
Ordering information: Supplied with a one-year manufacturer’s warranty on parts. Glove box can be equipped as anaerobic chambers with the addition of digital catalyst heater units; contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty