Would you like to log in again?
TipStation Pipette Tip Systems, Sorenson BioScience
Supplier: Sorenson BioScience
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TipStation
27760T 27770T 27790 27790T
87000-772CS 312.39 USD InStock
87000-772 87000-762 87000-770 87000-764
TipStation Pipette Tip Systems, Sorenson BioScience
Pipette Tips
Stackable racks of tips are ideal for use with 8- and 12-channel pipettors.
  • Racks, lids, and base are manufactured from recycled plastic
Racks snap together and will not lift during pipetting. Recessed wells keep the tips stable for mounting on pipettors. The stacked design saves bench space and features a wide base to prevent the stack from tipping.
Ordering information: Each rack contains 192 tips in two 8×12 trays. The stack includes five racks of tips, one lid, and one base.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty