Would you like to log in again?
Poly (l-lysine hydrobromide)
Supplier: Polysciences
09730-25
87001-894EA 165.95 USD InStock
87001-894
Poly (l-lysine hydrobromide)
Proteins and Peptides
Cationic polymer
Used to improve cell adhesion to solid surfaces. 0.1% soln. in water.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty