Would you like to log in again?
Fluoresbrite® YG Microspheres, Calibration Grade 6 µm, Polysciences
Supplier: Polysciences
Fluoresbrite®
18862-1
87001-906EA 1317.11 USD InStock
87001-906
Fluoresbrite® YG Microspheres, Calibration Grade 6 µm, Polysciences
Controls and Calibration Verification Materials
For flow cytometry standards
A special grade of Yellow Green Fluoresbrite® beads has been evaluated for both particle diameter and uniformity of dye distribution. Packaged in an economical suspension of 2.5% solids in water
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty