Would you like to log in again?
Poly (D,L-lactide-co-glycolide), 50:50, IV 1.0 dl/g, Polysciences
Supplier: Polysciences
23987-5
87001-914EA 502.39 USD InStock
87001-914
Poly (D,L-lactide-co-glycolide), 50:50, IV 1.0 dl/g, Polysciences
Media for Microbiology Additives and Supplements for Microbiology
Poly (DL-lactide-co-glycolide) (PLGA) is a well-established biodegradable polymer used in medical devices and drug delivery applications
In comparison to Poly (L-lactide) (PLLA), PLGA offers control over degredation rate and reduced glass transition temperatures (by increasing glycolide content). The control of these properties makes PLGA especially well suited for applications where medical devices are only needed for weeks (e.g. drug delivery).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty