Would you like to log in again?
Tissue Capsules, Simport
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M470 M470 M471 M471
87002-380CS 384.84 USD InStock
87002-380 87002-378 CA73960-010 CA73960-012
Tissue Capsules, Simport
Embedding Cassettes Tissue Capsules
These polypropylene capsules are suitable for holding tissue samples during processing
The lids have a frosted marking area for sample identification and an open mesh area to facilitate fluid exchange. The whole surface of the base is also an open mesh. The lid snaps securely on the base, eliminating the risk of tissue loss during processing. Color: white.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty