Would you like to log in again?
Disposable Base Molds, Simport Scientific
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M475-4 M475-3 M475-3 M475-4 M475-5 M475-2 M475-10 M475-1 M475-5 M475-1 M475-2 M475-10
60872-490CS 305.36 USD InStock
CA60872-018 60872-490 60872-500 CA60872-014 60872-488 87002-474 CA60872-016 CA60872-020 60872-498 CA60872-023 CA60872-022 60872-486
Disposable Base Molds, Simport Scientific
Moulds Moulds for Histology
These base molds offer the ease and convenience of a disposable mold, yet they are strong enough to be reused.
The specially formulated plastic material offers excellent thermal exchange. The molds have a smooth interior finish and rounded corners to facilitate specimen removal. They can be used with any standard cassettes and embedding rings.
The M475-10 base mold is designed especially for the M512 MACROSETTE®.
Delivery information: Each case contains two dispenser boxes of 500 base molds.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty