Would you like to log in again?
Storage Drawer, Simport Scientific
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M495-7 M495-7
87002-598CS 184.09 USD InStock
87002-598 CA73960-101
Storage Drawer, Simport Scientific
Boxes Storage Boxes
Made of durable, waterproof, heavy-duty cardboard for long-term storage.
  • Requires minimal space
  • Each drawer can store up to 165 embedding rings or 250 cassettes
Label provided for each drawer allows recording of specimen number, date, and type of specimens.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty