Would you like to log in again?
ProteCol™ HPLC Unions, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
ProteCol™
213006 212007 211010 223006 223005 222005 212011 222011 223007 213007 222007 212010 222009 212008 222006 212009 222010
87003-092EA 44.62 USD InStock
87003-092 87003-070 10799-702 87003-082 87003-094 87003-072 87003-074 87003-030 87003-032 87003-078 75815-598 87003-024 75815-600 87003-026 87003-028 75815-596 87003-090
ProteCol™ HPLC Unions, Trajan Scientific and Medical
Fittings and Connectors
SGE HPLC Unions are specifically designed and precision engineered for use in capillary LC
  • Zero dead volume design for no peak dispersion
  • Easy to use
  • Complete versatility
  • Allows connection to the same or different sized tubing
  • Available in PEEK and stainless steel
Stainless steel unions can be finger tightened or tightened with a 3/16" wrench for high-pressure applications. PEEK unions can be finger tightened. They are slightly larger than stainless steel unions, but they are lighter for less stress on your tubing. Pressure rating is 5000 psi for the stainless steel unions and 2175 psi for the PEEK unions.
ProteCol™ Unions and reusable PEEK ferrules allow you to connect any combination of 1/32" PEEKsil™, 1/16" PEEKsil and 0.36 mm fused silica tubing. Use for connecting guard or trap columns to analytical columns, or for bypassing the analytical column.
The ideal way to avoid any void or dead volume in a connection is to achieve a faultless butt connection
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: ProteCol unions are available in either stainless steel or PEEK (for all sizes). All use the same range of reusable PEEK ferrules.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty