Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrofluoric acid 47 - 51%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®

Supplier: VWR International
Danger

20502-VWNF05 20502-VWNF07 20502-VWNF03
87003-221EA 1893.47 USD InStock
87003-221 87003-222 87003-223
Hydrofluoric acid 47 - 51%, ARISTAR® ULTRA, Ultrapure for trace metal analysis, VWR Chemicals BDH®
Hydrofluoric acid
Ultra high purity acid for quantitative trace metal analysis at the parts-per-trillion (ppt) level.


Formula: HF
MW: 20.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,II
Order Now
Specification Test Results
Assay (HF, w/w) 47 - 51%
Total Sulfur (S) 100 ppb (ng/g) max.
Aluminum (Al) 20 ppt (pg/g) max.
Antimony (Sb) 20 ppt (pg/g) max.
Arsenic (As) 50 ppt (pg/g) max.
Barium (Ba) 10 ppt (pg/g) max.
Beryllium (Be) 10 ppt (pg/g) max.
Bismuth (Bi) 10 ppt (pg/g) max.
Boron (B) 100 ppt (pg/g) max.
Cadmium (Cd) 10 ppt (pg/g) max.
Calcium (Ca) 10 ppt (pg/g) max.
Cerium (Ce) 10 ppt (pg/g) max.
Cesium (Cs) 10 ppt (pg/g) max.
Chromium (Cr) 10 ppt (pg/g) max.
Cobalt (Co) 10 ppt (pg/g) max.
Copper (Cu) 10 ppt (pg/g) max.
Dysprosium (Dy) 1 ppt (pg/g) max.
Erbium (Er) 1 ppt (pg/g) max.
Europium (Eu) 1 ppt (pg/g) max.
Gadolinium (Gd) 1 ppt (pg/g) max.
Gallium (Ga) 10 ppt (pg/g) max.
Germanium (Ge) 10 ppt (pg/g) max.
Gold (Au) 20 ppt (pg/g) max.
Hafnium (Hf) 10 ppt (pg/g) max.
Holmium (Ho) 1 ppt (pg/g) max.
Indium (In) 1 ppt (pg/g) max.
Iron (Fe) 10 ppt (pg/g) max.
Lanthanum (La) 10 ppt (pg/g) max.
Lead (Pb) 10 ppt (pg/g) max.
Lithium (Li) 10 ppt (pg/g) max.
Lutetium (Lu) 1 ppt (pg/g) max.
Magnesium (Mg) 10 ppt (pg/g) max.
Manganese (Mn) 10 ppt (pg/g) max.
Mercury (Hg) 50 ppt (pg/g) max.
Molybdenum (Mo) 10 ppt (pg/g) max.
Neodymium (Nd) 1 ppt (pg/g) max.
Nickel (Ni) 20 ppt (pg/g) max.
Niobium (Nb) 10 ppt (pg/g) max.
Palladium (Pd) 20 ppt (pg/g) max.
Platinum (Pt) 20 ppt (pg/g) max.
Potassium (K) 10 ppt (pg/g) max.
Praseodymium (Pr) 1 ppt (pg/g) max.
Rhenium (Re) 10 ppt (pg/g) max.
Rhodium (Rh) 20 ppt (pg/g) max.
Rubidium (Rb) 20 ppt (pg/g) max.
Ruthenium (Ru) 20 ppt (pg/g) max.
Samarium (Sm) 1 ppt (pg/g) max.
Scandium (Sc) 10 ppt (pg/g) max.
Selenium (Se) Information only
Silver (Ag) 10 ppt (pg/g) max.
Sodium (Na) 10 ppt (pg/g) max.
Strontium (Sr) 10 ppt (pg/g) max.
Tantalum (Ta) Information only
Tellurium (Te) 1 ppt (pg/g) max.
Terbium (Tb) 1 ppt (pg/g) max.
Thallium (Tl) 10 ppt (pg/g) max.
Thorium (Th) 1 ppt (pg/g) max.
Thulium (Tm) 1 ppt (pg/g) max.
Tin (Sn) 20 ppt (pg/g) max.
Titanium (Ti) 20 ppt (pg/g) max.
Tungsten (W) 20 ppt (pg/g) max.
Uranium (U) 1 ppt (pg/g) max.
Vanadium (V) 10 ppt (pg/g) max.
Ytterbium (Yb) 1 ppt (pg/g) max.
Yttrium (Y) 1 ppt (pg/g) max.
Zinc (Zn) 10 ppt (pg/g) max.
Zirconium (Zr) 10 ppt (pg/g) max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty