Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 34 - 37%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®

Supplier: VWR International

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Muriatic acid

10401-VWNQ09 10401-VWNQ09 10401-VWNQ03
87003-251EA 168.02 USD InStock
87003-251 87003-253 CA87003-253
Hydrochloric acid 34 - 37%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Hydrochloric acid
High purity acid for quantitative trace metal analysis at the parts-per-billion (ppb) level.


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 34 - 37 %
Color max. 10 APHA
Aluminum (Al) max. 1 ppb
Antimony (Sb) max. 0.5 ppb
Arsenic (As) max. 0.5 ppb
Barium (Ba) max. 0.1 ppb
Beryllium (Be) max. 0.1 ppb
Bismuth (Bi) max. 0.1 ppb
Boron (B) max. 1 ppb
Cadmium (Cd) max. 0.1 ppb
Calcium (Ca) max. 1 ppb
Cerium (Ce) max. 0.1 ppb
Cesium (Cs) max. 0.1 ppb
Chromium (Cr) max. 0.5 ppb
Cobalt (Co) max. 0.1 ppb
Copper (Cu) max. 0.5 ppb
Dysprosium (Dy) max. 0.1 ppb
Erbium (Er) max. 0.1 ppb
Europium (Eu) max. 0.1 ppb
Gadolinium (Gd) max. 0.1 ppb
Gallium (Ga) max. 0.1 ppb
Gold (Au) max. 0.5 ppb
Hafnium (Hf) max. 0.1 ppb
Holmium (Ho) max. 0.1 ppb
Indium (In) max. 0.1 ppb
Iron (Fe) max. 1 ppb
Lanthanum (La) max. 0.1 ppb
Lead (Pb) max. 0.1 ppb
Lithium (Li) max. 0.1 ppb
Lutetium (Lu) max. 0.1 ppb
Magnesium (Mg) max. 0.5 ppb
Manganese (Mn) max. 0.1 ppb
Mercury (Hg) max. 0.1 ppb
Molybdenum (Mo) max. 0.1 ppb
Neodymium (Nd) max. 0.1 ppb
Nickel (Ni) max. 0.5 ppb
Niobium (Nb) max. 0.1 ppb
Palladium (Pd) Information Only
Platinum (Pt) Information Only
Potassium (K) max. 1 ppb
Praseodymium (Pr) max. 0.1 ppb
Rhenium (Re) max. 0.1 ppb
Rhodium (Rh) max. 0.1 ppb
Rubidium (Rb) max. 0.1 ppb
Ruthenium (Ru) max. 0.1 ppb
Samarium (Sm) max. 0.1 ppb
Scandium (Sc) max. 0.1 ppb
Selenium (Se) max. 1 ppb
Silver (Ag) max. 1 ppb
Sodium (Na) max. 1 ppb
Strontium (Sr) max. 0.1 ppb
Tantalum (Ta) Information Only
Tellurium (Te) max. 0.1 ppb
Terbium (Tb) max. 0.1 ppb
Thallium (Tl) max. 0.1 ppb
Thorium (Th) max. 0.1 ppb
Thulium (Tm) max. 0.1 ppb
Tin (Sn) max. 0.5 ppb
Titanium (Ti) max. 0.5 ppb
Tungsten (W) max. 0.1 ppb
Uranium (U) max. 0.1 ppb
Vanadium (V) max. 0.5 ppb
Ytterbium (Yb) max. 0.1 ppb
Yttrium (Y) max. 0.1 ppb
Zinc (Zn) max. 1 ppb
Zirconium (Zr) max. 0.1 ppb
Bromide (Br-) max. 10 ppm
Free Chlorine (Cl2) max. 0.5 ppm
Total Phosphorus (P) max. 0.01 ppm
Total Sulfur (S) max. 0.3 ppm

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty