Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Microcentrifuge Tubes, PP

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


VWR®
3310-E 3315 3300-E 3309 3305 3303-E 3307 3298 3304-E 3299 3297 3316 3312-E 3306 3311-E 3314 3308 3313-E 3302 3296-E 3301
87003-290CS 1056.8 USD InStock
87003-290 87004-268 87003-292 87004-258 87003-294 87004-266 87004-256 87004-278 87004-264 87003-296 87004-276 87004-254 87003-298 87004-262 87004-274 87004-260 87004-282 87003-300 87004-272 87004-280 87004-270
VWR® Microcentrifuge Tubes, PP
Tubes Microcentrifuge Tubes
Polypropylene tubes feature superior clarity and a positive seal design that withstands repeated cap closures and will not pop open during boiling.

  • Secure positive seal design
  • Tubes fit all standard rotors
  • Certified RNase-, DNase-, and pyrogen-free
  • Certified PCR inhibitor- and human DNA-free
  • Autoclavable

Temperature range is from –80 to 121 °C (–112 to 250 °F).

Ordering information: Rainbow packs consist of red, blue, green, yellow, and purple tubes.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty