Would you like to log in again?
Agarose, Molecular Biology Grade, IBI Scientific
Supplier: IBI Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
IB70035 IB70042 IB70040 IB70041 IB70047 IB70045 IB70046 IB70042
87004-980EA 1025.79 USD InStock
87004-980 IB70042 95043-304 97060-748 95000-010 IB70040 490007-708 97060-746
Agarose, Molecular Biology Grade, IBI Scientific
Electrophoresis Gels Agarose and Acrylamide
IBI Molecular Biology agarose products are highly refined and highly purified
Some features of IBI agarose include extraordinary mechanical resistance which insures safe and easy handling, exceptionally low absorption of staining reagents, excellent transparency which allows for high visibility, and greater thermal stability due to high hysteresis.
IBI agarose can be used for Nucleic Acid Electrophoresis, Analytical Electrophoresis, Preparative Electrophoresis, Blotting, and Protein Electrophoresis as in Radial Immunodiffusion.
Each lot is physically tested and verified for performance in its' designated application. IBI applies strict testing parameters to insure lot to lot consistency. IBI agarose products are offered in a wide variety of package sizes to fit the needs of any research. Each bottle of IBI agarose comes packaged with a specification sheet, preparation instructions, and MSDS for your convenience.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty