Would you like to log in again?
Agarose, 3:1 Super Sieve, IBI Scientific
Supplier: IBI Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
IB70052 IB70053
87004-984EA 857.93 USD InStock
87004-984 87004-982
Agarose, 3:1 Super Sieve, IBI Scientific
Electrophoresis Gels Agarose and Acrylamide
3:1 Super-Sieve Agarose is designed to have a larger gel network than IBI Ultra Sieve Agarose and is recommended for use in the separation of DNA fragments smaller than 1500bp
Some features of 3:1 Super-Sieve Agarose include excellent DNA resolution for sharp and finely resolved bands, very low background after EtBr staining, good transference for blotting of DNA fragments 154-2176bp, and very low DNA binding.
The range of separation for DNA bands in a 2.0% gel is 500-1500 bp. The range of separation for DNA in a 4.0% gel is 150-600bp. These ranges are an approximate and were calculated in 1X TAE Buffer.
Gels made with IBI 3:1 Super-Sieve Agarose have a higher gel strength than most competitive products. The gel is exceptionally firm, but still flexible when handled, thus minimizing cracking. IBI 3:1 Super-Sieve Agarose has the same melting and gelling temperature as our molecular biology grade agarose, therefore allowing fast and easy preparation of gels. The 3:1 Super-Sieve Agarose is recommended for all analytical applications, especially when DNA is recovered for subsequent use in enzymatic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty