Would you like to log in again?
ESR-Chex Controls, Streck
Supplier: Streck
214104 214112 214108 214102
87005-300BX 119.77 USD InStock
87005-300 87005-198 87005-200 87005-304
ESR-Chex Controls, Streck
Control reagents
Two-level hematology controls designed to monitor erythrocyte sedimentation rates
Manufactured from human red blood cells, these controls verify the precision and accuracy of both manual and automated methods. ESR-Chex is assayed for the most frequently used sedimentation rate tubes in conjunction with Classical Westergren, Modified Westergren, and Wintrobe methods. The removal of sodium citrate is not required for automated and modified procedures. ESR-Chex is to be used in the same manner as a patient sample. It has an open-vial stability for 95 days at room temperature, and a closed-vial stability for one year at 2 to 10°C (36 to 50°F).
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty