Would you like to log in again?
A1c-Cellular® Whole Blood Cellular Controls, Streck
Supplier: Streck
A1c-Cellular®
211124 211130
87005-434BX 327.58 USD InStock
87005-434 87005-428
A1c-Cellular® Whole Blood Cellular Controls, Streck
Control reagents
Controls test the entire HbA1c procedure, incluing the lysing of the red blood cell, which ensures the entire system, instrument and reagents are working properly
The controls have an open-vial stability of 30 days, and a closed-vial stability of 6 months when stored at 2 to 10°C. Available in plastic vials with pierceable caps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty