Would you like to log in again?
Infectious Disease Tests and Controls, SA Scientific™
Supplier: SA Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
069101 611530 041130 041101 067101 069020
87007-030PK 248.35 USD InStock
87007-030 87007-020 87007-022 87007-026 87007-028 87007-018
Infectious Disease Tests and Controls, SA Scientific™
Clinical Diagnostic Supplies Clinical Reagents and Kits
These rapid diagnostic tests are designed to facilitate the detection of a variety of viral and bacterial infectious diseases.
Three viral infection tests are available: adenovirus, which can detect all 41 known adenovirus strains; rotavirus, which can detect all rotavirus strains; and RSV, which can detect the respiratory syncytial virus. Controls to validate results must be purchased separately. Tests are conducted by adding specimen samples into sample wells and simply reading results, which are available within 15 minutes.
The bacterial infection StrepAlert™ test provides an immunoassay for Strep A. Controls are included with this test. The procedure is conducted with a dipstick and provides results within five minutes.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty