Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Disposable Microcentrifuge Tubes

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
CFT458005 CFT457020 CFT451015 CFT450015 CFT454015 CFT458020 CFT458015 CFT455020 CFT450020 CFT454005 CFT456005 CFT451020 CFT452015 CFT455015 CFT454020 CFT452020 CFT453005 CFT450005 CFT453020 CFT455005 CFT456020 CFT451005 CFT452005 CFT457005 CFT456015 CFT457015 CFT453015
89000-040CS 330.42 USD InStock
89000-040 89000-028 20170-084 20170-088 20170-086 89000-020 89000-042 89000-044 89000-022 89000-024 89000-026 89000-030 20170-094 89000-018 20170-170 20170-092 20170-098 20170-096 89000-032 89000-010 89000-012 89000-034 89000-036 20170-090 89000-014 89000-038 89000-016
VWR® Disposable Microcentrifuge Tubes
Tubes Microcentrifuge Tubes
These tubes are made from USP Class VI PP.

  • Amber colored for light-sensitive substances
  • RNase-, DNase-, and endotoxin-free
  • Engraved graduations ensure accuracy
  • Caps can be penetrated by a syringe and will not pop open during centrifuging

These tubes also feature flat, pierceable, frosted caps and a side labeling surface.

Autoclavable at 121 °C and freezable to –80 °C.

Certifications: Manufactured in a Class 100,000 cleanroom environment. Manufactured under ISO13485:2016 and ISO9001:2015 quality management systems. Meet the requirements of RoHS.

Ordering information: Assorted pack contains 100 tubes each of red, orange, yellow, green and blue.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty