Would you like to log in again?
VWR® Sterile Sample Bags
Supplier: Avantor
VWR®
EFR-1015-VW1 EPR-7012-VW1 EPL-3070-VW1 BPL-4512-VW1 EPL-5515-VW1 BFR-5590-VW1 EFL-4590-VW1 EPR-7012-VW1 EDL41214-VW1 EPL-4590-VW1 EPR-4590-VW1 EPR47012-VW1 EFL4712S-VW1 EDL41216-VW1 BFR-4590-VW1 EFR-5590-VW1 EFL47012-VW1 EPL-7012-VW1 EPR-3070-VW1 EDL41216-VW1 EPL45590-VW1 EFL-1015-VW1 EFL47012S-VW1 BPL-5515-VW1 SCL-2424-VW1 EPL-4512-VW1 SCL33030-VW1 BFL-5515-VW1 EFR-1012-VW1 EPL-5590-VW1 BFL-3070-VW1 BPR-3050-VW1 EPL-4590-VW1 EFR-1015-VW1 BFL-5590-VW1 EPR47012-VW1 EDL41212-VW1 EDL41218-VW1 BPL-7012-VW1 EPR-4590-VW1 EPL-3070-VW1 EFR-1012-VW1 EFL-5590-VW1 EPL47012-VW1 BPR-5590-VW1 BPR-7012-VW1 EFR-4590-VW1 SCL33037-VW1 EPR-5590-VW1 EFR47012-VW1 EFL-1012-VW1 BPR-7012-VW1 EPL-5590-VW1 BFL45590-VW1 SCL21520-VW1 EDL41212-VW1 BPR-4590-VW1 SCL21520-VW1 EPL-4575-VW1 BPL-5590-VW1 EPL-5515-VW1 EPR-3050-VW1 BPL-5590-VW1 BPR-5590-VW1 BPL-4512-VW1 BFL-5590-VW1 BFL-5515-VW1 BFL45590-VW1 EFR-3070-VW1 EPR-5590-VW1 EDL41224 EPR-7012S-VW1 EPL-7012-VW1 EFL-1012-VW1 EPR-4512-VW1 EPR-3070-VW1 EDL41214-VW1 BPL-7012-VW1 EPL-3050-VW1 EDL41218-VW1 EDL41224-VW1 BFL-4512-VW1 EFR-5590-VW1 BPR-3070-VW1 BPL-4590-VW1 EFL-1015-VW1 BFR-5590-VW1 EFR47012-VW1 EFL-5515-VW1 EFL-4590-VW1 BPR-4590-VW1 EPL45590-VW1 EFR-4590-VW1 BFR-3070-VW1 EDL41524-VW1 EPL47012-VW1 BFL-4512-VW1 EPR45590-VW1 BPL-5515-VW1 EPR-3050-VW1 EPL-7015-VW1 EFL-5590-VW1 EPL-3070-VW1
89085-556CS 403.89 USD InStock
89085-556 89004-418 89085-558 89085-552 CA89085-560 89085-554 11216-104 89000-644 89000-648 CA11216-104 89004-420 89085-550 89004-422 CA89000-650 89000-640 89004-424 89000-642 82007-696 CA89004-418 89000-190 82007-698 CA89000-642 89000-636 CA89000-644 89000-638 CA89000-648 CA89004-420 89085-560 CA89004-422 CA89000-640 CA89004-424 CA89085-542 89085-535 89085-530 CA89085-540 CA89000-190 89085-532 89085-538 89085-537 CA89085-546 CA89000-636 CA89000-638 CA89085-556 89085-544 89085-546 89085-540 76214-710 CA89085-552 CA89085-550 89085-542 89085-548 CA89085-558 10818-386 76221-334 CA83007-556 82007-700 76289-492 CA89085-528 CA82007-710 82007-708 CA83007-558 CA82007-716 82007-706 CA82007-714 CA82007-718 83007-556 CA89085-532 83007-558 CA83007-560 CA89085-530 CA83007-562 CA82007-702 CA82007-700 CA82007-706 CA89085-538 CA82007-704 89085-528 CA82007-708 83007-560 CA82007-694 CA82007-692 76196-376 83007-562 CA82007-698 CA82007-696 76196-372 82007-726 82007-724 82007-728 76265-604 CA89085-580 76196-370 CA82007-720 CA82007-724 82007-714 CA82007-722 CA82007-728 82007-718 CA82007-726 82007-716 89085-580 89000-650 CA82007-690
VWR® Sterile Sample Bags
Bags Sample Bags
These sample bags are ideal for transportation and storage of solids, semisolids, and liquids for environmental and carcass sampling, biomedical and pharmaceutical research, quality assurance procedures, food industry applications, and clinical and veterinary medicine.
  • Pliant polyethylene sterile tubing
  • Highly resistant
  • Wire closure for an airtight seal
  • Round wire bags have two wires for closure
Bags are made of pliant, highly resistant virgin polyethylene sterile tubing with no side seals. They feature a wire closure for an airtight seal, and wide-sealed bottoms for added leakproof protection. To use, tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
Note: Do not use with alcoholic substances or where temperature exceeds 100 °C (212 °F).
Certifications: Comply with EPA, FDA, USDA, and HACCP regulatory and industry requirements.
Ordering information: Bags 89085-560 and 76289-492 are packaged in a double pouch for cleanroom applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty