Would you like to log in again?
WypAll® X70 Wipers, Kimberly-Clark Professional®
Supplier: Kimberly-Clark
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WYPALL®
41600 41200 41702 41100 41600 41061 41455 41600 41061 41100 41702 41200 41455 41412 55300 41611 41611
89003-020CS 213.89 USD InStock
89003-020 37002-040 37002-052 89003-012 CA89003-020 89003-016 CA89003-012 CA89003-018 89003-018 CA89003-016 89000-688 CA89000-688 CA37002-028 CA21907-032 37002-028 CA37002-052 76325-982
WypAll® X70 Wipers, Kimberly-Clark Professional®
Wipes Reusable Cleaning Cloths Cloths for General Applications
WypAll® X70 Cloths are durable, reusable, absorbent, and stay strong for better cleaning performance.
  • Fast absorbing HYDROKNIT® material is a hydroentangled cellulose/polypropylene blend with no adhesives or chemical binders
  • Durable, rag-tough construction and textured surface
  • Reusable, will not fall apart when wet
  • Ideal for cleaning oil, dirt, grime, and solvents
  • Picks up water almost instantly, and hangs on for fewer drips, outperforming laundered shop rags
For use in heavy manufacturing, transportation fleet maintenance, metal fabrication, machine shops, tool and die shops, and processing plants for chemicals, refining, rubber, and plastics. Made of fast absorbing HYDROKNIT™ material, a hydroentangled cellulose/polypropylene blend without adhesives or chemical binders. Feature durable, rag-tough construction and a textured surface, yet are soft and pliable. Wipers can stand up to the toughest industrial cleaning tasks. They are reusable and will not fall apart when wet. Long-lasting, on-the-job performance allows extended usage, reducing cost and waste.
Pound for pound absorbs more oil and water than rags.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty