Would you like to log in again?
Polyethylene [ST] Stoppers, Kimble Chase
Supplier: DWK Life Sciences
28160R-13 28160R-27 28160R-16 28160R-9 28160R-22 28160R-19
89000-799CS 36.8 USD InStock
89000-799 89000-801 89000-803 89000-805 89000-807 89000-797
Polyethylene [ST] Stoppers, Kimble Chase
Stoppers PE/LDPE/HDPE Stoppers
For items ground to [ST] stopper dimensions
Stoppers are yellow and blue with a closed bottom. Made from linear high density polyethylene. Enlarged flange is designed to protect the neck if the glassware item is tipped over.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty