Would you like to log in again?
KIMAX® Filling Funnels, Short Stem, 58° Angle, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
28950-55 28950-90 28950-100 28950-75 28950-45 28950-25 28950-35 28950-65
89001-410CS 596.38 USD InStock
89001-410 89000-826 89000-828 89001-408 89001-406 89001-404 89001-412 89001-402
KIMAX® Filling Funnels, Short Stem, 58° Angle, Kimble Chase, DWK Life Sciences
Funnels Filtration Funnels Filter Funnels
Constructed with a strong, molded rim, heavy uniform walls, and a fire-polished stem for a long service life.
Unique 58° funnel angle provides faster and more effective filtration than the standard 60° funnel design. Funnels also have a high resistance to chemical attack and mechanical and thermal shock.
Certifications: Designed from ASTM Specification E1095, Type 1, Class A requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty