Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
PTFE Key-Head [ST] Stoppers, Flask Length Joint, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
41901R-9 41901R-16 41901R-32 41901R-27 41901R-13 41901R-22 41901R-8 41901R-19 41901R-38
89000-832CS 103.84 USD InStock
89000-832 89000-834 10135-810 89000-836 89000-838 16358-236 16358-238 10135-808 89000-830
PTFE Key-Head [ST] Stoppers, Flask Length Joint, Kimble Chase
Stoppers PE/LDPE/HDPE Stoppers
Stoppers are for use with flasks, mixing cylinders, separatory funnels, and more
Designed from ASTM Specification E675.
Color-coded handles indicate stopper size. Stoppers size 9 through 22 are solid. Sizes 32 and 38 are hollow.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty