Would you like to log in again?
Single Metric Scale Cylinders with Bumper, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
20024-01
89000-968CS 280.77 USD InStock
89000-968
Single Metric Scale Cylinders with Bumper, Kimble Chase
Cylinders Measuring Cylinders
An assortment of five popular sized graduated cylinders (one each) in sizes of 10, 25, 50, 100 and 250 mL
The 10 mL cylinder has graduations from 1 to 10 mL; the 25 mL cylinder has graduations from 3 to 25 mL; the 50 mL cylinder has graduations from 3 to 50 mL; the 100 mL cylinder has graduations from 5 to 100 mL; the 250 mL cylinder has graduations from 10 to 250 mL.
Ideal for start-up requirements and customers who need a variety of cylinders, but have limited lab space or glassware needs.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty