Would you like to log in again?
KIMAX® ColorWare Erlenmeyer Flasks, Narrow Mouth, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
26500G-1000 26500B-1000 26500R-250 26500R-1000 26500Y-1000 26500G-50 26500B-50 26500Y-50 26500B-125 26500R-125 26500Y-500 26500R-50 26500G-250 26500Y-250 26500R-500 26500G-125 26500G-500 26500B-250 26500B-500 26500Y-125
89001-014CS 119.16 USD InStock
89001-014 89001-004 89001-026 89001-034 89001-012 89001-024 89001-002 89001-032 89001-010 89001-022 89001-030 89001-020 89001-018 89001-008 89001-016 89001-006 89001-028 89000-996 89000-998 89000-994
KIMAX® ColorWare Erlenmeyer Flasks, Narrow Mouth, Kimble Chase
Flasks Erlenmeyer Flasks
Available with markings in four colors, the KIMAX® ColorWare flasks are brighter in appearance than traditional glassware with white markings.
All flasks have durable, colored, ceramic enamel scales to indicate approximate volumes at various levels. Designed from ASTM Specification E1404, Type I, Class I requirements. Manufactured from 33 expansion, low extractable borosilicate glass, conforming to USP Type I and ASTM E438, Type I, Class A requirements.
These flasks are also commonly used to help reduce cross-contamination and for easy identification of glassware between labs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty