Would you like to log in again?
KIMAX® Graduated Cylinders, Class B, To Contain, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
20022-250 20022-500 20022-50 20022-25 20022-100 20022-10 20022-2000 20022-1000
89001-102CS 402.09 USD InStock
89001-102 89001-100 89001-098 89001-096 89001-094 89001-108 89001-106 89001-104
KIMAX® Graduated Cylinders, Class B, To Contain, Kimble Chase
Cylinders Measuring Cylinders
Designed from ASTM E1272, Style 1, Class B requirements. The 10mL size has funnel top and sizes 25mL and larger have SAFE-GARD® bumpers.
  • Calibrated to contain at 20°C
  • Single metric scale is durable white ceramic enamel
  • Hexagonal base provides stability
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty