Would you like to log in again?
KIMAX® Graduated Cylinders, Class B, To Deliver, with Red Stripe, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
20024D-50 20024D-2000 20024D-250 20024D-100 20024D-25 20024D-500 20024D-10 20024D-1000
89001-136CS 966.51 USD InStock
89001-136 89001-134 89001-132 89001-140 89001-130 89001-128 89001-126 89001-138
KIMAX® Graduated Cylinders, Class B, To Deliver, with Red Stripe, Kimble Chase
Cylinders Measuring Cylinders
Cylinders are made of sturdy borosilicate glass
Designed from ASTM Specification E1272, Style 1, Class B requirements.
A clear, reverse image in the single metric scale is highlighted against a ruby red stripe. The 10mL cylinder has an enlarged funnel-shaped top for ease of filling. Cylinders 25mL and larger are supplied with SAFE-GARD® bumpers, which are sturdy split rings made of yellow low-density polyethylene.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty