Would you like to log in again?
KIMAX® Graduated Cylinders, Class B, To Deliver, Blue Scale, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
20025-1000 20025-25 20025-100 20025-500 20025-2000 20025-10 20025-50 20025-250
89001-146CS 1012.49 USD InStock
89001-146 89001-144 89001-156 89001-142 89001-154 89001-152 89001-150 89001-148
KIMAX® Graduated Cylinders, Class B, To Deliver, Blue Scale, Kimble Chase
Cylinders Measuring Cylinders
Printed with durable blue enamel ceramic single metric scale
Designed from ASTM Specification E1272, Style 1, Class B requirements.
Cylinders with 25 through 2000mL capacities are supplied with a SAFE-GARD® bumper. The 10mL size has an enlarged funnel-shaped top for ease of filling.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty