Would you like to log in again?
KIMAX® Culture Flasks, Capacity Scale, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
25615-1000 25615-2000 25615-125 25615-250 25615-500
89001-300CS 1554.92 USD InStock
89001-300 89001-298 89001-296 89000-780 89001-302
KIMAX® Culture Flasks, Capacity Scale, Kimble Chase
Flasks Cell Culture Flasks
Flasks have a thick-walled, long tubing neck with a plain fire-polished finish sealed to an Erlenmeyer-shaped body
Designed from ASTM Specification E1404, Type IV, Class 1 requirements.
Neck is long enough to accept a metal or plastic cap or a cotton plug. Durable white ceramic enamel scale indicates approximate volume.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty