Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KIMBLE® KIMAX® Gooch Crucibles, Low Form, with KIMFLOW® Fritted Disc, DWK Life Sciences

Supplier: DWK Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

KIMAX®
28260-303 28260-302 28260-152 28260-301
89001-366CS 79.57 USD InStock
89001-366 89001-370 89000-808 89001-368
KIMBLE® KIMAX® Gooch Crucibles, Low Form, with KIMFLOW® Fritted Disc, DWK Life Sciences
Crucibles
Especially suitable for precipitates to be dried to constant weight at 110°C

Note: Exposure to sudden temperature changes should be avoided.

For higher temperatures, an electric furnace is advised and crucibles should not be removed until temperature has dropped to 250°C.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty