Would you like to log in again?
KIMAX® Culture Tubes, Reusable, with Screw Cap, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
45066-13100 45066-20125 45066-16150 45066-25200 45066-16100 45066-38200 45066A-20125 45066-16125 45066A-13100 45066A-16125 45066A-20150 45066A-25200 45066A-16100 45066-25150 45066-20150 45066A-16150 45066A-25150
89001-476CS 586.54 USD InStock
89001-476 89001-466 89001-488 89001-474 89001-464 89001-486 89001-472 89001-462 89001-484 89001-470 89001-482 89001-478 89001-468 89001-490 89001-480 89000-860 89000-862
KIMAX® Culture Tubes, Reusable, with Screw Cap, Kimble Chase
Tubes Culture Tubes
Tubes with uniform wall thickness for maximum heat transfer and chemical resistance.
Tubes feature a marking spot and a threaded phenolic screw cap that can be loosened for autoclaving. Tubes are available with rubber-lined caps or PTFE-faced rubber-lined caps.
Certifications: Tubes with rubber-lined caps are designed from ASTM Specification E982, Type VI, Class A requirements. Tubes with PTFE-faced rubber-lined caps are designed from ASTM Specification E982, Type VI, Class B requirements.
Ordering information: Cap is supplied unattached.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty