Would you like to log in again?
Phenolic Screw Thread Closure, Kimble Chase
Supplier: DWK Life Sciences
45066C-24 73804-20400 45066B-18 73804-15425 45066B-28 45066C-38 45066B-15 45066C-18 45066C-13 73804-18400 45066B-38 73804-24400 45066C-15 45066B-13 45066B-24
89001-498CS 843.6 USD InStock
89001-498 89001-048 66010-694 66010-672 66010-683 89001-496 89001-494 89001-492 89001-506 66010-887 89001-504 89001-502 89001-500 16197-302 89001-050
Phenolic Screw Thread Closure, Kimble Chase
Closures Test/Sample Tube Closures
Caps have mineral-filled phenolic material to withstand the effects of repeated autoclaving.
Rubber-lined caps retain adhesive properties and meet ASTM Specification E982, Class A requirements. PTFE-faced rubber-lined caps meet ASTM specification E982, Class B requirements.
Unlined closures have excellent chemical resistance and are autoclavable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty