Would you like to log in again?
KIMAX® Test Tubes, Reusable, Borosilicate Glass, with Glass Stopper, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
45100-16150 45100-19150 45100-25200 45100-13100
89000-864CS 1243.86 USD InStock
89000-864 89001-510 89000-866 89001-512
KIMAX® Test Tubes, Reusable, Borosilicate Glass, with Glass Stopper, Kimble Chase
Tubes Culture Tubes
Test tubes made from KG-33 borosilicate glass with a [ST] top and a [ST] glass stopper.
Uniform wall thickness for maximum service. Tubes incorporate a durable white ceramic marking spot. Designed from ASTM Specification E982, Type III requirements.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty