Would you like to log in again?
KIMBLE® KIMAX® Reservoir Bottles with Bottom Hose Outlet, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
KIMAX®
14607-5000 14607-20000 14607-2000 14607-1000 14607-500 14607-250 14607-10000
89001-532CS 184.61 USD InStock
89001-532 89001-540 89001-530 89001-528 89001-538 89001-536 89001-534
KIMBLE® KIMAX® Reservoir Bottles with Bottom Hose Outlet, DWK Life Sciences
Bottles Aspirators
Reservoir bottles feature a narrow mouth, tooled top, and white enamel marking spot.
Glass hose connection outlet is fused to the bottle. Sizes 250 and 500 ml accept 7.9mm (⁵/₁₆") I.D. flexible tubing; 1000 ml and larger sizes accept 9.5 mm (³/₈") Int. Ø tubing.
Certifications: Borosilicate glass conforms to ASTM E438, Type I, Class A.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty