Would you like to log in again?
KIMAX® Heavy-Duty Solution Bottles, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
14950-500 14950-120 14950-35 14950-25
89001-552CS 573.67 USD InStock
89001-552 89001-550 89003-472 89001-548
KIMAX® Heavy-Duty Solution Bottles, Kimble Chase, DWK Life Sciences
Bottles Solution Bottles
Borosilicate glass carboy-style bottles feature a narrow neck and sloping shoulders.
Necks are tooled for a uniform fit with a #12 rubber stopper (not included). The 19 L (5 gal.) size conforms to Federal Specification DD-B-597.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty