Would you like to log in again?
KIMAX® Dropping Bottles, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
15035-15 15035-30 15035-125 15035-60
89001-564CS 708.87 USD InStock
89001-564 89001-562 89001-560 89001-566
KIMAX® Dropping Bottles, Kimble®, DWK Life Sciences
Bottles Dropping and Dispensing Bottles
These borosilicate glass bottles have 1 ml dropper capacity and include a medium length [ST] pipet dropper with a rubber nipple.
The 30 and 60 ml sizes are KG-33 ASTM 438, Type 1, Class A glass and meet Federal Specification NNN-B-1194, Type II, Class 2, Grade B requirements. The 125 ml size is KG-35 ASTM, 438 Type 1, Class B glass.
Bottles are suitable for use in determining the oil absorption of pigments (ASTM D281).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty