Would you like to log in again?
KIMBLE® KIMAX® Gas Washing Bottles, Tall Form, [ST] Stopper, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
15060-125 15060-250 15060-500
89001-572CS 391.21 USD InStock
89001-572 89001-570 89001-568
KIMBLE® KIMAX® Gas Washing Bottles, Tall Form, [ST] Stopper, DWK Life Sciences
Bottles Gas Washing Bottles
For washing or drying gases.
Gas flows into the bottom of the bottle through the center tube, rises through the washing or drying medium, and exits through the side tube of the stopper. Bottles have [ST] 34/28 stoppers and inlet and outlet ports that accept 0.63 cm (¹/₄") Int.Ø flexible tubing.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty