Would you like to log in again?
KIMAX® Gas Washing Bottle, Short Form, [ST] Stopper, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
15065-250
89001-574CS 302.86 USD InStock
89001-574
KIMAX® Gas Washing Bottle, Short Form, [ST] Stopper, DWK Life Sciences
Bottles Gas Washing Bottles
For washing or drying gases.
  • Shorter and wider
  • Bottle with medium length joint
  • Manufactured from 33 expansion, low extractable borosilicate glass
Gas flows into the bottom of the bottle through the center tube, rises through the washing or drying medium, and exits through the side tube of the stopper. Bottles have inlet and outlet ports that accept ¹/₄" Int.Ø flexible tubing.
Certifications: Conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty