Would you like to log in again?
KIMAX® Solution Bottles with Color-Coded PTFE [ST] Flathead Stopper, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
15097-100 15097-2000 15097-250 15097-500 15097-1000
89001-576CS 99.68 USD InStock
89001-576 89001-584 89001-582 89001-580 89001-578
KIMAX® Solution Bottles with Color-Coded PTFE [ST] Flathead Stopper, Kimble Chase, DWK Life Sciences
Bottles Solution Bottles
Narrow-mouth bottles conform to ASTM E438, Type 1, Class A standards.
Bottle necks are [ST] ground to accept flathead PTFE stoppers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty