Would you like to log in again?
KIMAX® Medium Length West Condenser, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
18185-1420
89001-660CS 327.32 USD InStock
89001-660
KIMAX® Medium Length West Condenser, Kimble Chase
Condensers
KIMAX® 200mm (77/8") one-piece sealed unit with a [ST] 14/20 outer joint at the inlet and a [ST] 14/20 inner joint at the outlet
Inlet end is sloped to avoid trapping condensate. Inner tube is light-walled with a small diameter jacket to keep minimal space between them. Hose connections accept 7.9mm (5/16") I.D. flexible tubing. (Kimble Chase 18185 1420)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty