Would you like to log in again?
KIMAX® Graduated Cylinders, Class A, To Deliver, Serialized and Certified, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
20026-100 20026-1000 20026-10 20026-50 20026-25 20026-500 20026-250
89001-674CS 897.24 USD InStock
89001-674 89001-672 89001-670 89001-678 89001-668 89001-676 89001-666
KIMAX® Graduated Cylinders, Class A, To Deliver, Serialized and Certified, Kimble Chase
Cylinders Measuring Cylinders
Cylinders are made of sturdy borosilicate glass with a hexagonal glass base that has been flat ground for stability.
Cylinders feature a single metric scale
Designed from ASTM Specification E1272, Style 1, Class A requirements. Each cylinder is permanently marked with an individual serial number and is supplied with a Certificate of Graduation Accuracy.
The 10mL cylinder has an enlarged funnel-shaped top for ease of filling. Cylinders 25mL and larger are supplied with SAFE-GARD® bumpers, which are sturdy split rings made of yellow low-density polyethylene.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty