Would you like to log in again?
KIMAX® Reverse Scale Graduated Cylinders, Class A, To Deliver, Kimble Chase
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KIMAX®
20028W-1000 20028W-2000 20028W-10 20028W-250 20028W-50 20028W-500 20028W-100 20028W-25
89001-696CS 446.96 USD InStock
89001-696 89001-686 89001-694 89001-684 89001-692 89001-682 89001-690 89001-688
KIMAX® Reverse Scale Graduated Cylinders, Class A, To Deliver, Kimble Chase
Cylinders Measuring Cylinders
Cylinders are made of sturdy borosilicate glass and feature reverse metric graduations in white
Cylinders 25mL and larger are supplied with SAFE-GARD® bumpers, which are sturdy split rings made of yellow low-density polyethylene.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty