Would you like to log in again?
KIMAX® Graduated Cylinders, Class A, To Contain, Serialized and Certified, with Glass Stoppers, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
20036-50 20036-10 20036-500 20036-25 20036-100 20036-250 20036-1000
89001-708CS 1427.84 USD InStock
89001-708 89001-706 89001-718 89001-716 89001-714 89001-712 89001-710
KIMAX® Graduated Cylinders, Class A, To Contain, Serialized and Certified, with Glass Stoppers, Kimble Chase
Cylinders Measuring Cylinders
Cylinders and [ST] stoppers are made of sturdy borosilicate glass
Cylinders are designed from ASTM Specification E1272, Style 2, Class A requirements. Each cylinder is permanently marked with an individual serial number and is supplied with a Certificate of Graduation Accuracy. Stoppers are designed from ASTM Specification E675.
Cylinders are suitable for mixing. Single metric scale is in durable white ceramic enamel.
Ordering information: Replacement stoppers are available; see the 89002-174 series.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty