Would you like to log in again?
KIMAX® Volumetric Flasks with Snap Cap, Class A, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
28008-1000 28008-100 28008-200 28008-250 28008-500 28008-50 28008-10 28008-25 28008-2000
89001-862CS 494.24 USD InStock
89001-862 89001-860 89001-858 89001-856 89001-868 89001-854 89001-866 89001-852 89001-864
KIMAX® Volumetric Flasks with Snap Cap, Class A, Kimble Chase
Flasks Volumetric Flasks
Calibrated to contain
Designed from ASTM Specification E288 and with Class A accuracy but without serial number.
Supplied with a polyethylene snap cap.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty