Would you like to log in again?
KIMAX® Volumetric Flasks with Snap Cap, Class B, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
28010-100 28010-250 28010-50 28010-2000 28010-500 28010-200 28010-25 28010-1000 28010-10
89001-872CS 508.48 USD InStock
89001-872 89001-884 89001-870 89001-882 89001-880 89001-878 89001-876 89001-874 89001-886
KIMAX® Volumetric Flasks with Snap Cap, Class B, Kimble Chase
Flasks Volumetric Flasks
Calibrated to contain
Designed from ASTM Specification E288, Class B requirements.
Flasks have a graduation ring blasted on the neck and a marking spot. Neck opening is tooled for the supplied polyethylene snap cap. The enlarged top of cap protects the neck if flask is tipped over.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty