Would you like to log in again?
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class B, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
28015-1000 28015-2000 28015-250 28015-100 28015-500 28015-50 28015-200 28015-25 28015-10
89001-994CS 308.34 USD InStock
89001-994 89002-002 89001-992 89001-990 89001-988 89001-986 89001-998 89001-984 89001-996
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class B, Kimble Chase
Flasks Volumetric Flasks
Calibrated to contain
Designed from ASTM specification E288, Class B requirements.
Flasks have a graduation ring blasted on the neck, a marking spot, and a [ST] ground glass stopper.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty