Would you like to log in again?
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class A, Serialized and Certified, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
28017-25 28017-500 28017-200 28017-250 28017-10 28017-100 28017-1000 28017-50 28017-2000
89002-034CS 1127.58 USD InStock
89002-034 89002-024 89002-036 89002-040 89002-030 89002-032 89002-026 89002-038 89002-028
KIMAX® Volumetric Flasks with [ST] Glass Stopper, Class A, Serialized and Certified, Kimble Chase
Flasks Volumetric Flasks
Calibrated to contain
Designed from ASTM Specification E288, Class A serialized requirements. Flasks meet requirements of NBS Circular 602 and come with a Certificate of Graduation Accuracy.
Flasks are marked with individual serial numbers. Flasks have a graduation ring blasted on neck, a marking spot, and a [ST] ground glass stopper.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty