Would you like to log in again?
KIMAX® Micro Volumetric Flasks with [ST] Glass Stopper, Cylindrical, Class A, Serialized and Certified, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
28017A-25 28017A-5 28017A-10 28017A-2 28017A-1
89002-044CS 599.67 USD InStock
89002-044 89002-046 89002-042 89002-050 89002-048
KIMAX® Micro Volumetric Flasks with [ST] Glass Stopper, Cylindrical, Class A, Serialized and Certified, Kimble Chase
Flasks Volumetric Flasks
Calibrated to contain
Designed from ASTM Specification E237, Class A serialized requirements.
Flasks have individual serial numbers, a marking spot, and a graduation ring blasted on the neck. Cylindrical body is ideal for better mixing and drainage and for ease in withdrawal of samples using pipets. Sizes 1, 2, and 5mL have a circular base, and 10 and 25mL sizes have a hexagonal base. Supplied with a Certificate of Graduation Accuracy and [ST] ground glass stopper.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty