Would you like to log in again?
KIMAX® Separatory Funnel, Globe, with PTFE Stopper, DWK Lifesciences
Supplier: DWK Life Sciences
KIMAX®
29043F-250 29043F-125
89002-084CS 269.78 USD InStock
89002-084 89002-086
KIMAX® Separatory Funnel, Globe, with PTFE Stopper, DWK Lifesciences
Funnels Separating Funnels
The sharp separations in these funnels are provided for by a narrow bore in the body just above the stopcock.
  • Gradually tapering shoulders and a wide neck facilitate cleaning
  • Designed from ASTM Specification E1096, Type 2 requirements
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Delivery information: Includes standard taper ground glass stopper and a PTFE stopcock.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty